2-Isocyanato-1-methoxy-4-methylbenzene 98%
Catalog # APOSOR480677-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:163.18 g/mol
Boiling Pt:240 °C/760mmHg
Melting Pt:25…26 °C
Density:1.112 g/cm³ (25 °C)
Storage Temperature:Freezer
UN:2206
ADR:6.1,III
CAS Number:59741-04-7
Formula:C₉H₉NO₂
MDL Number:MFCD00013869