Order Entry
Netherlands
ContactUsLinkComponent
2-(2-Bromoethyl)-1,3-dioxane 98%
Irritant
2-(2-Bromoethyl)-1,3-dioxane 98%
Catalog # APOSOR480586-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(2-Bromoethyl)-1,3-dioxane 98%
Catalog # APOSOR480586-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :195.06 g/mol
Boiling Pt:67…70 °C
Density:1.431 g/cm³ (25 °C)
Flash Pt:97 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:33884-43-4
Formula:C₆H₁₁BrO₂
MDL Number:MFCD00006567