Order Entry
Netherlands
ContactUsLinkComponent
1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne
1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne
Catalog # APOSOR470909-1G
Supplier:  Apollo Scientific
CAS Number:  
1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyne
Catalog # APOSOR470909-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :277.96 g/mol
Storage Temperature:Ambient
CAS Number:1010840-17-1
Formula:C₁₄H₂₄B₂O₄
MDL Number:MFCD22666429