Tetrahydropyran-4-sulphonyl chloride
Catalog # APOSOR460012-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.634 g/mol
Storage Temperature:Ambient
CAS Number:338453-21-7
Formula:C₅H₉ClO₃S
MDL Number:MFCD07787616