Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
Ethyl-2-iodobenzoate
Catalog # APOSOR4535-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-2-iodobenzoate
Catalog # APOSOR4535-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :276.07 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1829-28-3
Formula:C₉H₉IO₂
MDL Number:MFCD00051796