N-Cyclopentyltetrahydro-2H-pyran-4-amine hydrochloride
Catalog # APOSOR451046-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.73 g/mol
Storage Temperature:Ambient
CAS Number:859521-03-2
Formula:C₁₀H₂₀ClNO
MDL Number:MFCD11035935