
2-(Chloromethyl)-6-methylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3099-29-4
:
Formula:C₇H₈ClN
MDL Number:MFCD09055262
:141.6000 g/mol
Boiling Pt:72…75 °C
Melting Pt:18…19 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together