Order Entry
Netherlands
ContactUsLinkComponent
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaboranlan-2-yl)phenyl]-1H-pyrazole
Irritant
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaboranlan-2-yl)phenyl]-1H-pyrazole
Catalog # APOSOR43580-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaboranlan-2-yl)phenyl]-1H-pyrazole
Catalog # APOSOR43580-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :270.14 g/mol
Melting Pt:75…76 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:852227-94-2
Formula:C₁₅H₁₉BN₂O₂
MDL Number:MFCD07368520