Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
Methyl-4-bromo-1H-pyrazole-3-carboxylate
Catalog # APOSOR42123-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-4-bromo-1H-pyrazole-3-carboxylate
Catalog # APOSOR42123-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.0094 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:81190-89-8
Formula:C₅H₅BrN₂O₂
MDL Number:MFCD00466326