
5-Bromo-2-methoxy-4-methyl-3-nitropyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:884495-14-1
:
Formula:C₇H₇BrN₂O₃
MDL Number:MFCD06659504
:247.0500 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together