Order Entry
Netherlands
ContactUsLinkComponent
3-Hydroxy-1-methacryloyloxyadamantane 97%
3-Hydroxy-1-methacryloyloxyadamantane 97%
Catalog # APOSOR41028-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Hydroxy-1-methacryloyloxyadamantane 97%
Catalog # APOSOR41028-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :236.31 g/mol
Storage Temperature:Ambient
CAS Number:115372-36-6
Formula:C₁₄H₂₀O₃
MDL Number:MFCD13152044