Order Entry
Netherlands
ContactUsLinkComponent
Methyl-2-chlorobenzoate 99+%
Irritant
Methyl-2-chlorobenzoate 99+%
Catalog # APOSOR4068-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-2-chlorobenzoate 99+%
Catalog # APOSOR4068-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:233…235 °C
Density:1.191 g/cm³ (25 °C)
Flash Pt:108 °C
 :170.6 g/mol
EINECS:210-242-0
 :GHS07
Storage Temperature:Ambient
CAS Number:610-96-8
Formula:C₈H₇ClO₂
MDL Number:MFCD00016337