2-(Pyridin-4-yl)ethane-1-sulphonyl chloride
Catalog # APOSOR400473-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.66 g/mol
Storage Temperature:Ambient
CAS Number:252670-82-9
Formula:C₇H₈ClNO₂S
MDL Number:MFCD08445803