
(2-Methyl-1H-imidazol-4-yl)methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:45533-87-7
:
Formula:C₅H₈N₂O
MDL Number:MFCD06202901
:112.13 g/mol
Melting Pt:120…122 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together