Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl-4-{2-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperazine-1-carboxylate 95+%
tert-Butyl-4-{2-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperazine-1-carboxylate 95+%
Catalog # APOSOR361624-250MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-{2-[4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)phenoxy]ethyl}piperazine-1-carboxylate 95+%
Catalog # APOSOR361624-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :432.37 g/mol
Storage Temperature:Ambient
CAS Number:1310404-00-2
Formula:C₂₃H₃₇BN₂O₅
MDL Number:MFCD12827855