1-Phenylsulphonyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Catalog # APOSOR361535-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:334.20 g/mol
Storage Temperature:Ambient
CAS Number:1073372-04-9
Formula:C₁₅H₁₉BN₂O₄S
MDL Number:MFCD09037504