2-Methoxy-3-(methoxycarbonyl)-5-pyridineboronic acid pinacol ester 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1083168-93-7
:
Formula:C₁₄H₂₀BNO₅
MDL Number:MFCD11855973
:293.1233 g/mol
Storage Temperature:Kamertemperatuur
Frequently Bought Together