Order Entry
Netherlands
ContactUsLinkComponent
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Catalog # APOSOR361070-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester 97%
Catalog # APOSOR361070-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :262.16 g/mol
Storage Temperature:Ambient
CAS Number:1233526-60-7
Formula:C₁₄H₂₃BN₂O₂
MDL Number:MFCD13176540