
2-Methoxy-4-methyl-5-pyridineboronic acid
Catalog # APOSOR360255-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:166.97 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:503184-35-8
Formula:C₇H₁₀BNO₃
MDL Number:MFCD07368880