
3-Hydrazinylbenzonitrile hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2881-99-4
:
Formula:C7H8ClN3
MDL Number:MFCD03094642
Melting Pt:194…196 °C
:169.6115 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together