Order Entry
Netherlands
ContactUsLinkComponent
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride
Catalog # APOSOR350392-100MG
Supplier:  Apollo Scientific
CAS Number:  
5,6,7,8-Tetrahydro-1,6-naphthyridine hydrochloride
Catalog # APOSOR350392-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :170.64 g/mol
Storage Temperature:Ambient
CAS Number:1187830-51-8
Formula:C₈H₁₁ClN₂
MDL Number:MFCD11846187