Order Entry
Netherlands
ContactUsLinkComponent
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol
Catalog # APOSOR350354-5G
Supplier:  Apollo Scientific
CAS Number:  
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol
Catalog # APOSOR350354-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :171.2600 g/mol
Storage Temperature:Ambient
CAS Number:116539-55-0
Formula:C₈H₁₃NOS
MDL Number:MFCD07357308