Bestelling invoeren
Netherlands
ContactUsLinkComponent
(±)-α-Bromophenylacetic acid
(±)-α-Bromophenylacetic acid
Catalog # APOSOR350067-100G
Supplier:  Apollo Scientific
CAS Number:  
(±)-α-Bromophenylacetic acid
Catalog # APOSOR350067-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :215.0440 g/mol
Melting Pt:82…83 °C
Storage Temperature:Ambient
Storage Temperature:Kamertemperatuur
UN:3261
ADR:8,II
 :GHS05
 :GHS07
CAS Number:4870-65-9
Formula:C₈H₇BrO₂
MDL Number:MFCD00004206