Order Entry
Netherlands
ContactUsLinkComponent
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester
Irritant
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester
Catalog # APOSOR346186-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester
Catalog # APOSOR346186-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :156.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:51985-95-6
Formula:C₆H₈N₂O₃
MDL Number:MFCD01851055