N'-[1-Cyclopentylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1053657-97-8
:
Formula:C₁₈H₂₇N₃O₃
MDL Number:MFCD10568261
:333.43 g/mol
Storage Temperature:Kamertemperatuur
Frequently Bought Together