N'-[2-(4-Chlorophenyl)-1-[(furan-2-ylmethyl)amino]ethylidene]hydrazinecarboxylic acid tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1053657-85-4
:
Formula:C₁₈H₂₂ClN₃O₃
MDL Number:MFCD10568259
:363.85 g/mol
Storage Temperature:Ambient
Frequently Bought Together