N'-[1-Cyclobutylamino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1053657-19-4
:
Formula:C₁₇H₂₅N₃O₃
MDL Number:MFCD10568244
:319.41 g/mol
Storage Temperature:Ambient
Frequently Bought Together