5-(4-Bromophenyl)-4-methyl-2,4-dihydro-3H-1,2,4-triazole-3-thione 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:494194-50-2
:
Formula:C₉H₈BrN₃S
MDL Number:MFCD03995808
:270.16 g/mol
Storage Temperature:Ambient
Frequently Bought Together