Order Entry
Netherlands
ContactUsLinkComponent
2-(5-Methyl-1H-pyrazol-3-yl)pyrazine 95%
2-(5-Methyl-1H-pyrazol-3-yl)pyrazine 95%
Catalog # APOSOR32946-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(5-Methyl-1H-pyrazol-3-yl)pyrazine 95%
Catalog # APOSOR32946-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.18 g/mol
Storage Temperature:Ambient
CAS Number:192711-19-6
Formula:C₈H₈N₄
MDL Number:MFCD08056640