Order Entry
Netherlands
ContactUsLinkComponent
Methyl-2-(1H-1,2,4-triazol-1-yl)acetate 95%
Methyl-2-(1H-1,2,4-triazol-1-yl)acetate 95%
Catalog # APOSOR32778-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-2-(1H-1,2,4-triazol-1-yl)acetate 95%
Catalog # APOSOR32778-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :141.13 g/mol
Storage Temperature:Ambient
CAS Number:106535-16-4
Formula:C₅H₇N₃O₂
MDL Number:MFCD02186570