4-(3-Chlorophenyl)-4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine 95%
Catalog # APOSOR32457-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:233.7 g/mol
Storage Temperature:Ambient
CAS Number:18094-25-2
Formula:C₁₂H₁₂ClN₃
MDL Number:MFCD02102424