Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl-1-oxa-4,8-diazaspiro[4.5]decane-8-carboxylate 95%
 
tert-Butyl-1-oxa-4,8-diazaspiro[4.5]decane-8-carboxylate 95%
  APOSOR32277-250MG
 :  
 
tert-Butyl-1-oxa-4,8-diazaspiro[4.5]decane-8-carboxylate 95%
  APOSOR32277-250MG
 :  

 

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

 

 

 :
 :242.32 g/mol
 :GHS07
 :Ambient
 :886360-95-8
 :C₁₂H₂₂N₂O₃
 :MFCD07698579