Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
5-Piperazin-1-yl-1,6-naphthyridine hydrochloride 90%
Catalog # APOSOR318082-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Piperazin-1-yl-1,6-naphthyridine hydrochloride 90%
Catalog # APOSOR318082-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.73 g/mol
Melting Pt:>215 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:1171412-23-9
Formula:C₁₂H₁₅ClN₄
MDL Number:MFCD09753939
Melting Pt:>215 °C (dec.)