
6-Oxa-1-azaspiro[3.3]heptane hemioxalate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1380571-72-1
:
Formula:C₁₂H₂₀N₂O₆
MDL Number:MFCD22373634
:288.3 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together