
3-[(tert-Butyldimethylsilanyl)oxy]azetidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:875340-81-1
:
Formula:C₉H₂₁NOSi
MDL Number:MFCD20662828
:187.36 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together