Order Entry
Netherlands
ContactUsLinkComponent
6-(p-Toluenesulphonyl)-2-oxa-6-azaspiro[3.3]heptane
6-(p-Toluenesulphonyl)-2-oxa-6-azaspiro[3.3]heptane
Catalog # APOSOR317036-5G
Supplier:  Apollo Scientific
CAS Number:  
6-(p-Toluenesulphonyl)-2-oxa-6-azaspiro[3.3]heptane
Catalog # APOSOR317036-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :253.32 g/mol
Storage Temperature:Ambient
CAS Number:13573-28-9
Formula:C₁₂H₁₅NO₃S
MDL Number:MFCD13180735