
(R)-N-(tert-Butoxycarbonyl)-2-phenylglycinol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:102089-74-7
:
Formula:C₁₃H₁₉NO₃
MDL Number:MFCD00274205
:237.3 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together