Order Entry
Netherlands
ContactUsLinkComponent
trans-4-Aminoadamantan-1-ol hydrochloride
Irritant
trans-4-Aminoadamantan-1-ol hydrochloride
Catalog # APOSOR315778-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
trans-4-Aminoadamantan-1-ol hydrochloride
Catalog # APOSOR315778-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :203.7100 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:62075-23-4
Formula:C₁₀H₁₈ClNO
MDL Number:MFCD11099880