
tert-Butyl 2-oxo-6-azaspiro[3.4]octane-6-carboxylate
Catalog # APOSOR312268-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:225.284 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:203661-71-6
Formula:C₁₂H₁₉NO₃
MDL Number:MFCD17016202