
2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester
Catalog # APOSOR312267-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:239.32 g/mol
Melting Pt:121…123 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:203661-69-2
Formula:C₁₃H₂₁NO₃
MDL Number:MFCD12198552