
Boc-1-aminocyclohexane-1-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:115951-16-1
:
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD00273427
:243.31 g/mol
Melting Pt:177 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together