Order Entry
Netherlands
ContactUsLinkComponent
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride
Catalog # APOSOR310732-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[3-(3-Methyl-1,2,4-oxadiazol-5-yl)pyridin-2-yl]piperidin-4-amine dihydrochloride
Catalog # APOSOR310732-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :332.24 g/mol
Storage Temperature:Ambient
CAS Number:1239748-54-9
Formula:C₁₃H₁₉Cl₂N₅O
MDL Number:MFCD28023520