1-{[1-(2-Chlorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride
Catalog # APOSOR310731-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:350.68 g/mol
Storage Temperature:Ambient
CAS Number:1610377-07-5
Formula:C₁₃H₁₈Cl₃N₅
MDL Number:MFCD28023519