Ethyl-3-methylquinoxaline-2-carboxylate
Catalog # APOSOR310043-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.24 g/mol
Melting Pt:70…72 °C
Storage Temperature:Ambient
CAS Number:3885-38-9
Formula:C₁₂H₁₂N₂O₂
MDL Number:MFCD02182648