Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-7-methoxy-4-methylquinoline
2-Chloro-7-methoxy-4-methylquinoline
Catalog # APOSOR309383-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-7-methoxy-4-methylquinoline
Catalog # APOSOR309383-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :207.66 g/mol
Storage Temperature:Ambient
CAS Number:97892-67-6
Formula:C₁₁H₁₀ClNO
MDL Number:MFCD00711757