2-chloro-8-methylquinoline-3-carbaldehyde
Catalog # APOSOR309277-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Chloro-8-methylquinoline-3-carboxaldehyde, 2-Chloro-3-formyl-8-methylquinoline, 2-Chloro-8-methylquinoline-3-carbaldehyde
:
:205.641 g/mol
Storage Temperature:Ambient
CAS Number:73568-26-0
Formula:C₁₁H₈ClNO
MDL Number:MFCD02227041