(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:138108-72-2
:
Formula:C₁₀H₁₉NO₃
MDL Number:MFCD03094725
:201.27 g/mol
Storage Temperature:Ambient
Frequently Bought Together