Order Entry
Netherlands
ContactUsLinkComponent
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester
Catalog # APOSOR308116-1G
Supplier:  Apollo Scientific
CAS Number:  
(6-Oxospiro[3.3]hept-2-yl)carbamicacid-tert-butyl ester
Catalog # APOSOR308116-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.29 g/mol
Storage Temperature:Ambient
CAS Number:1118786-86-9
Formula:C₁₂H₁₉NO₃
MDL Number:MFCD11858160