


3-Bromophenyl isothiocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2131-59-1
:
Formula:C₇H₄BrNS
MDL Number:MFCD00004803
:214.09 g/mol
Boiling Pt:256 °C
Flash Pt:113 °C
UN:3335
ADR:9,III
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together