
tert-Butyl N-(azetidin-3-yl)-N-methylcarbamate hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:943060-59-1
:
Formula:C₉H₁₉ClN₂O₂
MDL Number:MFCD12963843
:222.712 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together